Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Class A Lifetime Red Volumetric Flask with Glass Standard Taper Stopper
GREENER_CHOICE

Catalog No. p-126580 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1 L
2000 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
10-210-8B 50 mL
10-210-8A 25 mL
10-210-8C 100 mL
10-210-8D 200 mL
10-210-8E 250 mL
10-210-8G 1 L
10-210-8F 500 mL
10-210-8H 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 10-210-8B Supplier Corning™ Supplier No. 566050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sharp, clear graduation lines

The capacity mark is a fine line etched through a narrow red band into clear glass, making a filler unnecessary and giving high legibility, accuracy, and permanence. With standard taper stopper. Calibrated 'To Contain.'



For stopper only, see Cat. No. 7650.
TRUSTED_SUSTAINABILITY

Specifications

Calibration To Contain
Diameter (English) Outer 2 in.
Capacity (Metric) 50 mL
Material Glass, Borosilicate Glass
Height (English) 5.98 in.
Height (Metric) 15.2 cm
Neck Style Long
No. per Case 12/Cs.
No. per Pack 6/Pk.
Tolerance ±0.05 mL
Graduated Yes
Class A
Bottom Shape Flat
Show More Show Less
What are the advantages of using PYREX Low Actinic Glassware?

• Reduces light transmission to protect sensitive samples
• Ensures stability and precision in experiments and long-term storage
• Durable construction for reliable use in various laboratory applications

What applications are PYREX Low Actinic Glassware suitable for?

PYREX Low Actinic Glassware is ideal for:
• Chemistry
• Biology
• Pharmaceuticals
• Photochemistry experiments
• Any process involving chemicals or substances sensitive to light in the 3,000 to 5,000 Angstrom range

Why use PYREX Low Actinic Glassware?

It is designed to reduce light transmission, particularly UV and visible wavelengths, protecting light-sensitive samples such as chemicals, reagents, and biological materials from degradation caused by light exposure. This minimizes light-induced reactions, ensuring the stability and integrity of stored or processed substances.

What is PYREX Low Actinic Glassware made of?

PYREX Low Actinic Labware is made from Type I, Class A borosilicate glass with copper replacing the sodium in the glass composition structure, ensuring durability equal to the base glass.

What are the key features of PYREX Low Actinic Glassware?

• Non-pyrogenic
• Human DNA-free
• PCR inhibitor-free
• RNase-/DNase-free
• Made with USP Class VI Polypropylene (for plastic components)
• Provides protection for materials sensitive to light

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.