Learn More
Electron Microscopy Sciences Tripod Polisher Pyrex Insert
SDP
Catalog No. 50980876
Encompass
The Pyrex Insert for Wedge Polishing (#50116-60): is used with the Screw Clamp and Slotted L-Bracket for mounting sample for the TEM - For use with Tripod Polisher Model 590.
Catalog No. 50-980-876 Supplier Electron Microscopy Sciences Supplier No. 5011660
The Pyrex Insert for Wedge Polishing (#50116-60): is used with the Screw Clamp and Slotted L-Bracket for mounting sample for the TEM - For use with Tripod Polisher Model 590.