Learn More
Electron Microscopy Sciences Low Form Beakers, Gradations: 20-80 ml. Divisions: 10 ml. Diameter: 50 mm. Height: 72 mm., 1/EA
SDP
Catalog No. 502090593
Encompass_Preferred
Manufactured with uniform wall thicknesses, these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements. Gradations: 20-80 ml. Divisions: 10 ml . Diameter: 50 mm . Height: 72 mm.
Catalog No. 50-209-0593 Supplier Electron Microscopy Sciences Supplier No. 6096101
Manufactured with uniform wall thicknesses, these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements. Gradations: 20-80 ml. Divisions: 10 ml . Diameter: 50 mm . Height: 72 mm.