Learn More
Electron Microscopy Sciences Low Form Beakers, Gradations: 25-200 ml. Divisions: 25 ml. Diameter: 68 mm. Height: 90 mm., 1/EA
SDP
Supplier:  Electron Microscopy Sciences 6096103
Encompass_Preferred
Manufactured with uniform wall thicknesses, these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements. Has a double graduated scale to indicate approximate volume. Gradations: 25-200 ml. Divisions: 25 ml . Diameter: 68 mm . Height: 90 mm.
Catalog No. 50-209-0595