Learn More
Electron Microscopy Sciences Low Form Beakers, Gradations: 50-500 ml. Divisions: 50 ml. Diameter: 90 mm. Height: 124 mm., 1/EA
SDP
Catalog No. 502090597
Encompass_Preferred
Manufactured with uniform wall thicknesses, these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements. Has a double graduated scale to indicate approximate volume. Gradations: 50-500 ml. Divisions: 50 ml . Diameter: 90 mm . Height: 124 mm.
Catalog No. 50-209-0597 Supplier Electron Microscopy Sciences Supplier No. 6096105
Add to Cart
Add to Cart
Manufactured with uniform wall thicknesses, these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements. Has a double graduated scale to indicate approximate volume. Gradations: 50-500 ml. Divisions: 50 ml . Diameter: 90 mm . Height: 124 mm.