Learn More
Corning™ PYREX™ Microchemistry Kit Accessories - Vacuum Adapter with Hose Connection
Catalog No. 1130084 Shop All Corning Products
Change view
Click to view available options
Standard Taper Joint:
14/10 bottom inner
Catalog No. Standard Taper Joint
11-300-84 14/10 bottom inner
1 options
Catalog No. 11-300-84 Supplier Corning™ Supplier No. 894810
Replacement part for PYREX Microchemistry Kits
  • With standard taper 14/10 thread, standard taper 14/10 joint, and 10mm serrated tubulation
  • Arm angle: 105°
Order Info
For 11-300-71 and 11-300-74 Microchemistry Kits
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For PYREX™ microchemistry kit
Includes Hose connection, 10mm serrated tubulation
Standard Taper Joint 14/10 bottom inner
Product Type Vacuum Adapter