Learn More
Electron Microscopy Sciences Pyrex Plate, 9 cavity Pyrex pressed plate 
SDP
Catalog No. 50931198
Encompass_Preferred
A 9 cavity Pyrex pressed plate which offers a clear view for observation by transmitted light. The plate measures: 4"(L) x 3 3/8"(W) (100x85 mm). The cavity is 1/4"(6.4 mm) deep with a 7/8" (22 mm) opening.
Catalog No. 50-931-198 Supplier Electron Microscopy Sciences Supplier No. 7156301
A 9 cavity Pyrex pressed plate which offers a clear view for observation by transmitted light. The plate measures: 4"(L) x 3 3/8"(W) (100x85 mm). The cavity is 1/4"(6.4 mm) deep with a 7/8" (22 mm) opening.