Learn More
Corning™ PYREX™ Replacement Tip For 2116-LSO
GREENER_CHOICE
Catalog No. 10458 Shop All Corning Products
Change view
Click to view available options
No. per Case:
12/Cs.
36/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
10-458 36/Cs.
09-552-122 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-458 Supplier Corning™ Supplier No. 6406GTO
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
For use with for Pyrex 25mL and 50mL Economy Grade Burets (Nos. 2116-25 and 2116-50)
Replacement tips for 2116 style buret
Order Info
Can be used only with the PTFE Locking Stopcock (No. 2116-LSO)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) PTFE Locking Stopcock
Material Borosilicate Glass
No. per Pack 12/Pk.
No. per Case 36/Cs.
Type Buret Tip