Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Reusable Media Storage Bottles
SCI_ED

Catalog No. S14265 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
3.5 mL
50 mL
100 mL
150 mL
250 mL
500 mL
750 mL
1000 mL
2000 mL
5000 mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S14265 750 mL
S14255 100 mL
S14256 250 mL
S14257 500 mL
S14258 1000 mL
S14262 50 mL
S14259 2000 mL
S14264 150 mL
S14260 5000 mL
S14263 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Bottles larger than 2L should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.
Catalog No. S14265 Supplier Corning™ Supplier No. 1395750/EMD
This item is not returnable. View return policy
Bottles larger than 2L should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.

Ideal for media, buffers, aqueous and nonaqueous solutions

These heavy duty PYREX™ round bottles can be used for storage as well as mixing and sampling. They include a linerless, one-piece autoclavable orange polypropylene plug seal GL32 threaded cap and a drip-free pouring ring. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Bottles have permanent white enamel graduations and marking spots. Caution: DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.
  • PYREX™
  • GL threaded polypropylene cap
  • Drip-free pouring ring
  • Permanent, white marking spot and graduations

Order Info

Optional red melamine caps and PTFE rings are available for applications requiring temperatures to 180°C (356°F).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 750 mL
Capacity (Metric) 750 mL
For Use With (Application) Storage, mixing, sampling
Includes Drip-free pouring ring
Material Borosilicate Glass (Type I)
Sterility Non-sterile
Graduations 100 to 400 mL
Closure Included Yes
Shape Cylindrical
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.