Learn More
Harrick Plasma Pyrex Sample Tray for PDC-001,PDC-001-HP
SDP
Catalog No. NC0897694
Encompass_Preferred
The PDC-00T-P Pyrex Sample Tray is 5.5"W x 6.5"L x 0.125” thick and provides a horizontal surface for loading small samples and for batch processing. Use with the PDC-001 Expanded Plasma Cleaner or PDC-001-HP High Power Expanded Plasma Cleaner.
Catalog No. NC0897694 Supplier Harrick Plasma Supplier No. PDC00TP
The PDC-00T-P Pyrex Sample Tray is 5.5"W x 6.5"L x 0.125” thick and provides a horizontal surface for loading small samples and for batch processing. Use with the PDC-001 Expanded Plasma Cleaner or PDC-001-HP High Power Expanded Plasma Cleaner.