Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Standard Taper Polyethylene Stoppers
GREENER_CHOICE

Catalog No. 1452948 Shop All Corning Products
Change view
Click to view available options
Stopper Number:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
14-529-48 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-529-48 Supplier Corning™ Supplier No. 76248
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Design protects neck during event of accidental tipping

These blue and white stoppers are made with closed bottoms, of linear high-density polyethylene to conform to standard taper stopper dimensions. The three unit rings on the base provide an efficient seal. The improved design fully protects the neck in the event of accidental tipping. These stoppers may be used in place of the conventional standard taper flask stoppers.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Tapered Stopper
Stopper Number 8
Material HDPE
For Use With (Application) Protects the neck in the event of accidental tipping
Color Blue, White
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.