Learn More
Corning™ PYREX™ Three-Way Claisen Adapters
GREENER_CHOICE
Catalog No. 11850103 Shop All Corning Products
Change view
Click to view available options
Standard Taper Joint No.:
19/22 Inner Bottom, 19/22 Outer Top
Catalog No. Standard Taper Joint No.
11-850-103 19/22 Inner Bottom, 19/22 Outer Top
1 options
Catalog No. 11-850-103 Supplier Corning™ Supplier No. 905019
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
With 19/22 standard taper joints.
Cat. No. 9040-24 has outer standard taper joints at the upper end and on the parallel sidearm. With inner joint at the lower end. All joints on each tube are of the same size.
Cat. Nos. 9050-14 and 9050-19 have a three-way tube with two outer and one inner joint of the same size.
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 4.84 in.
Length (Metric) 123 mm
Material Borosilicate Glass
Product Type Connecting Adapter
Standard Taper Joint No. 19/22 Inner Bottom, 19/22 Outer Top