Learn More
Corning™ PYREX™ Tube Adapters with standard taper Joints at 105° Angle
GREENER_CHOICE
Catalog No. 153224 Shop All Corning Products
Change view
Click to view available options
Arm Angle:
105°
Catalog No. Arm Angle
15-322-4 105°
1 options
This item is not returnable. View return policy
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Catalog No. 15-322-4 Supplier Corning™ Supplier No. 894024
Add to Cart
Add to Cart
This item is not returnable. View return policy
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Two-way tube with inner and outer standard taper 24/40 joint connections at 105° angle
The arms on these PYREX™ tube adapters are approximately 105° apart. One end is equipped with an outer 24/40 Standard Taper joint and the other end has an inner 24/40 Standard Taper joint. Used as a receiver adapter.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Cone Size 24/40 Inner bottom, 24/40 outer top
Arm Angle 105°
Product Type Tube Adapter
WARNING: Cancer - www.P65Warnings.ca.gov