Learn More
Corning™ PYREX™ Tube Adapters with standard taper Joints at 75° Angle
GREENER_CHOICE
Catalog No. 153222 Shop All Corning Products
Change view
Click to view available options
Arm Angle:
75°
Catalog No. Arm Angle
15-322-2 75°
1 options
This item is not returnable. View return policy
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Catalog No. 15-322-2 Supplier Corning™ Supplier No. 892024
Add to Cart
Add to Cart
This item is not returnable. View return policy
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Two-way tube with standard taper 24/40 inner joints at both ends
The arms on these PYREX™ tube adapters are approximately 75° apart. Both ends are equipped with an inner 24/40 Standard Taper joints. Often used as a stillhead adapter.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Cone Size Two 24/40 Inner bottom
Arm Angle 75°
Product Type Tube Adapter
WARNING: Cancer - www.P65Warnings.ca.gov