Learn More
Corning™ PYREX™ VISTA™ Short Neck Round Bottom Boiling Flask, Standard Taper Joint
GSA_VAGREENER_CHOICE
Catalog No. p-3007060 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
07-250-082 250 mL
07-250-083 500 mL
07-250-084 1 L
07-250-085 2000 mL
07-250-081 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 07-250-082 Supplier Corning™ Supplier No. 70320250
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for great shock resistance
These round bottom boiling flasks have full length outer standard taper joints, but with shorter necks. They are manufactured from Code 7740 borosilicate glass. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance. The round bottom boiling flasks are designed to ASTM E-1403. Adapter Cat. Nos. 8800, 8820, and 8825 are used to connect flasks with different size joints, thus allowing numerous combinations for diverse assemblies.
Compliance
Designed to ASTM E-1403
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Neck Style Short
No. per Case 4/Cs.
No. per Pack 2/Pk.
Standard Taper Joint 24/40
Disposable Reusable
Bottom Shape Round