Learn More
Electron Microscopy Sciences Tripod Polisher Pyrex Wedge Polishing Rod
SDP
Catalog No. 50980879
Encompass
The Pyrex Wedge Polishing Rod (#50116-63): is a replacement part used with the model #50116-35 Wedge Polishing Mount. - For use with Tripod Polisher Model 590
Catalog No. 50-980-879 Supplier Electron Microscopy Sciences Supplier No. 5011663
The Pyrex Wedge Polishing Rod (#50116-63): is a replacement part used with the model #50116-35 Wedge Polishing Mount. - For use with Tripod Polisher Model 590