Learn More
Electron Microscopy Sciences Tripod Polisher Pyrex Wedge Polishing Stub
SDP
Catalog No. 50980880
Encompass
The Pyrex Wedge Polishing Stub (#50116-64): is mounted to a Pyrex Insert (#50116-60) to produce a similar effect as is accomplished with the model #50116-61. - For use with Tripod Polisher Model 590.
Catalog No. 50-980-880 Supplier Electron Microscopy Sciences Supplier No. 5011664
The Pyrex Wedge Polishing Stub (#50116-64): is mounted to a Pyrex Insert (#50116-60) to produce a similar effect as is accomplished with the model #50116-61. - For use with Tripod Polisher Model 590.