Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXPLUSR heavy wall filt1L
Catalog No. 502908833
Encompass
Flask features a protective PVC coating. Autoclavable (121 C) and resistant to thermal shock. Tubulation O.D. is 10 mm on sizes up through 1 000 mL. Tubulation is 12.5 mm O.D. on 2 000 and 4 000 mL sizes. The tubulation is not coated. Do not place over direct heat or flame. Do notAheat above 121 C moist heat or 110 C dry heat.
Catalog No. 50-290-8833 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS653401L6EA
Flask features a protective PVC coating. Autoclavable (121 C) and resistant to thermal shock. Tubulation O.D. is 10 mm on sizes up through 1 000 mL. Tubulation is 12.5 mm O.D. on 2 000 and 4 000 mL sizes. The tubulation is not coated. Do not place over direct heat or flame. Do notAheat above 121 C moist heat or 110 C dry heat.