Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXPLUSR volumetric flas
Catalog No. 502909086
Encompass
Flask features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121 C) and resistant to thermal shock.
Catalog No. 50-290-9086 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS6564025012EA
Flask features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121 C) and resistant to thermal shock.