Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR 200mL heavy wall ce
Catalog No. 502908767
Encompass
Particularly useful in handling sputum specimens. The bottle enables digestion shaking neutralizing and centrifuging to be performed in one container. The tight-fitting polypropylene screw cap with PTFE liner reduces the possibility of aerosol escape.
Catalog No. 50-290-8767 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS126120012EA
Particularly useful in handling sputum specimens. The bottle enables digestion shaking neutralizing and centrifuging to be performed in one container. The tight-fitting polypropylene screw cap with PTFE liner reduces the possibility of aerosol escape.