Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR baffled Erlenmeyer
Catalog No. 502905366
Encompass
Pyrex(R) baffled Erlenmeyer flasks feature a heavy tooled rim for maximum durability.Baffle-designed Delong flasks promote optimal and consistent SOLUTIONon agitation when used with flask shakers.Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.Includes a marking spot for convenience.
Catalog No. 50-290-5366 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS44505006EA
Pyrex(R) baffled Erlenmeyer flasks feature a heavy tooled rim for maximum durability.Baffle-designed Delong flasks promote optimal and consistent SOLUTIONon agitation when used with flask shakers.Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.Includes a marking spot for convenience.