Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR baffled shaker flas1EA
Catalog No. 502900227
Encompass
Pyrex(R) baffled shaker flasks are made of optically clear borosilicate glass.Delong-style Erlenmeyer flasks feature three baffle indents to provide greater agitation of SOLUTIONons to improve oxygen or gas transfer when used with rotary or reciprocating shakers.Heavy fire-polished long necks reduce splashing and accept two-position PP closures.Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.Includes a marking spot for convenience.
Catalog No. 50-290-0227 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS44442L1EA
Pyrex(R) baffled shaker flasks are made of optically clear borosilicate glass.Delong-style Erlenmeyer flasks feature three baffle indents to provide greater agitation of SOLUTIONons to improve oxygen or gas transfer when used with rotary or reciprocating shakers.Heavy fire-polished long necks reduce splashing and accept two-position PP closures.Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.Includes a marking spot for convenience.