Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR centrifuge tubes co
Catalog No. 502905706
Encompass
Graduation Range 0.1-15 ml Gradudation Interval 0.1 ml
Catalog No. 50-290-5706 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS80821512EA
Graduation Range 0.1-15 ml Gradudation Interval 0.1 ml