Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR centrifuge tubes wi
Catalog No. 502901374
Encompass
formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches chips and cracks. Always use with correct sleeve or holder.
Catalog No. 50-290-1374 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS80601512EA
formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches chips and cracks. Always use with correct sleeve or holder.