Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR centrifuge tubes wi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS80805012EA
Encompass
formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches chips and cracks. Always use with correct sleeve or holder.
Catalog No. 50-290-5394