Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR crystallizing dish
Catalog No. 502905625
Encompass
Pyrex(R) crystallizing dish is ideal for storage and crystallization.The reusable flat dishes are temperature resistant and can withstand repeated dry or wet sterilization.Reinforced fire-polished rim prevents chipping.
Catalog No. 50-290-5625 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS31401504EA
Pyrex(R) crystallizing dish is ideal for storage and crystallization.The reusable flat dishes are temperature resistant and can withstand repeated dry or wet sterilization.Reinforced fire-polished rim prevents chipping.