Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR crystallizing dish
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS31401254EA
Encompass
Pyrex(R) crystallizing dish is ideal for storage and crystallization.The reusable flat dishes are temperature resistant and can withstand repeated dry or wet sterilization.Reinforced fire-polished rim prevents chipping.
Catalog No. 50-290-5627