Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR culture tubes with
Catalog No. 502908682
Encompass
These Pyrex culture tubes are supplied with deep-form 18-415 GPI threaded phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of high temperatures and steam in autoclaving and are provided with a white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations.
Catalog No. 50-290-8682 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS98253848EA
These Pyrex culture tubes are supplied with deep-form 18-415 GPI threaded phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of high temperatures and steam in autoclaving and are provided with a white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations.