Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR culture tubes with
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS982516X288EA
Encompass
These Pyrex culture tubes are supplied with deep-form 18-415 GPI threaded phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of high temperatures and steam in autoclaving and are provided with a white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations.
Catalog No. 50-290-8892