Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR disposable culture
Catalog No. 502905964
Encompass
Round bottom. Tubes are made of borosilicate glass. Screw-cap tubes are supplied without caps. Designed for both tissue culture and bacteriological work.
Catalog No. 50-290-5964 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS99447161000EA
Round bottom. Tubes are made of borosilicate glass. Screw-cap tubes are supplied without caps. Designed for both tissue culture and bacteriological work.