Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXR Disposable Glass Co
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS9950215125EA
Encompass
These disposable 15 mL Pyrex(R) glass centrifuge tubes are manufactured from Type I borosilicate glass and have a GPI 15-415 thread. They are supplied without caps. The tubes are designed to withstand centrifugation up to 3 000 RCf and are packed in convenient easy to use packs which are shrink-sealed for protection during shipment. for reusable phenolic screw caps with PTfE(R) faced rubber liners see No. CLS999815 for disposable phenolic screw cap with rubber liner see No. CLS9999915.
Catalog No. 50-290-1324