Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXR Disposable Glass Co
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS995025072EA
Encompass
These disposable 50mL Pyrex glass centrifuge tubes are manufactured from Type I borosilicate glass and have a GPI 24-400 thread. They are supplied without caps. The tubes are designed to withstand centrifugation up to 3 000 RCf and are packed in convenient easy to use packs which are shrink-sealed for protection during shipment. for reusable phenolic screw caps with white rubber liners see CLS999924 for reusable phenolic screw caps with Polytetrafluorethylene (PTfE) faced rubber liners see CLS999824.
Catalog No. 50-290-6063