Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXR Disposable Glass Co
Catalog No. 502901382
Encompass
These disposable 10mL Pyrex glass centrifuge tubes are manufactured from Type I borosilicate glass and have a GPI 15-415 thread. They are supplied without caps. The tubes are designed to withstand centrifugation up to 3 000 RCf and are packed in convenient easy to use packs which are shrink-sealed for protection during shipment. for reusable phenolic screw caps with white rubber liners see CLS9999152 for reusable phenolic screw caps with Polytetrafluorethylene (PTfE) faced rubber liners see CLS999815 for disposable phenolic screw cap with rubber liner see CLS9999915.
Catalog No. 50-290-1382 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS9950210125EA
Add to Cart
Add to Cart
These disposable 10mL Pyrex glass centrifuge tubes are manufactured from Type I borosilicate glass and have a GPI 15-415 thread. They are supplied without caps. The tubes are designed to withstand centrifugation up to 3 000 RCf and are packed in convenient easy to use packs which are shrink-sealed for protection during shipment. for reusable phenolic screw caps with white rubber liners see CLS9999152 for reusable phenolic screw caps with Polytetrafluorethylene (PTfE) faced rubber liners see CLS999815 for disposable phenolic screw cap with rubber liner see CLS9999915.