Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR fernbach baffled cu
Supplier: Sigma Aldrich Fine Chemicals Biosciences CLS44232XL3EA
This 2800mL wide mouth PYREX(R) fernbach-style culture flask is designed for culturing organisms requiring a large surface area to volume ratio. It can also be used in serum production. Triple baffled on the bottom outside edges to achieve maximal oxygen transfer to culture medium. Beaded neck rim is approximately 60mm I.D. and accepts cotton plugs or No. 13 rubber stoppers. This flask has a large marking spot. Note Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.