Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR fernbach baffled cu
Catalog No. 502905837
Encompass
These 2800mL wide mouth PYREX(R) fernbach-style culture flasks have triple baffles located at the center of the flask bottom to achieve maximal oxygen transfer to culture medium. These triple baffled flasks have a Delong-style 38mm neck for use with plastic or stainless steel closures. for plastic closures see Nos. 26796-38 or 26797-38. These flasks have a large marking spot. Note Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.
Catalog No. 50-290-5837 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS44242XL3EA
These 2800mL wide mouth PYREX(R) fernbach-style culture flasks have triple baffles located at the center of the flask bottom to achieve maximal oxygen transfer to culture medium. These triple baffled flasks have a Delong-style 38mm neck for use with plastic or stainless steel closures. for plastic closures see Nos. 26796-38 or 26797-38. These flasks have a large marking spot. Note Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.