Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR fernbach culture fl
Catalog No. 502905839
Encompass
Wide-mouth design for culturing organisms requiring a large surface area to volume ratio. Can also be used in serum production. Reference Industrial and Engineering Chemistry Vol. 21 No. 12 1198. Note Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.
Catalog No. 50-290-5839 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS44202XL6EA
Wide-mouth design for culturing organisms requiring a large surface area to volume ratio. Can also be used in serum production. Reference Industrial and Engineering Chemistry Vol. 21 No. 12 1198. Note Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.