Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR gas dispersion tube
Catalog No. 502906091
Encompass
With 12Amm O.D. fritted cylinder 250Amm L x 8Amm diam. stem. Used to disperse gas in liquids. Size of bubbles depends on porosity. Size permits insertion through small openings.
Catalog No. 50-290-6091 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS3953312C6EA
With 12Amm O.D. fritted cylinder 250Amm L x 8Amm diam. stem. Used to disperse gas in liquids. Size of bubbles depends on porosity. Size permits insertion through small openings.