Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR Gooch crucible high
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS3294050C9EA
Encompass
Particularly adapted to analytical work where precipitates are dried to constant weight at 110C. Also suitable for higher temperatures in which case is advisable to heat in an electric furnace.Crucibles should not be subjected to sudden temperature changes.In order to avoid strain they should not be removed from the furnace until the temperature has dropped to 250C.
Catalog No. 50-290-5472