Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXR Griffin beakers low
Catalog No. 502900542
Encompass
Low form (Corning 1000 series) Griffin beakers have the following featuresGraduated for approximate contentDouble-graduated metric scaleBeakers have a spoutExtra-large area for markingUniform wall thicknessHeavy duty industrial grade glass beakers
Catalog No. 50-290-0542 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS10005012EA
Low form (Corning 1000 series) Griffin beakers have the following featuresGraduated for approximate contentDouble-graduated metric scaleBeakers have a spoutExtra-large area for markingUniform wall thicknessHeavy duty industrial grade glass beakers