Learn More
Sigma Aldrich Fine Chemicals Biosciences PYREXR Griffin beakers low
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS100010012EA
Encompass
Low form (Corning 1000 series) Griffin beakers have the following featuresGraduated for approximate contentDouble-graduated metric scaleBeakers have a spoutExtra-large area for markingUniform wall thicknessHeavy duty industrial grade glass beakers
Catalog No. 50-290-0849