Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR heavy wall drying t
Catalog No. 502908629
Encompass
These heavy duty Pyrex drying tray dishes will withstand hot air or steam sterilization and are useful for drying staining or developing trays. They can be used with gel electrophoresis or paper chromatography. Do not place over direct heat for oven use only. reusableheavy duty rim
Catalog No. 50-290-8629 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS3170124EA
These heavy duty Pyrex drying tray dishes will withstand hot air or steam sterilization and are useful for drying staining or developing trays. They can be used with gel electrophoresis or paper chromatography. Do not place over direct heat for oven use only. reusableheavy duty rim