Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR heavy wall graduate
Catalog No. 502914700
Encompass
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. for convenience the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Catalog No. 50-291-4700 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS53404L6EA
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. for convenience the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.