Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR heavy wall graduate1EA
Catalog No. 502905611
Encompass
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. for convenience the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Catalog No. 50-290-5611 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS53404L1EA
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. for convenience the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.