Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR Kjeldahl nitrogen d1EA
Catalog No. 502905819
Encompass
Used for determining nitrogen in organic compounds. This assembly includes a Kjeldahl flask specifically developed for this process. Both the digestion and distillation stages can be performed in this flask. the assembly shown is the distillation stage where the amount of ammonia and hence nitrogen is established. the inlet tube allows the permanganate SOLUTIONon to be introduced after distillation. Supplied with and outer ST 29/42 joint on the flask and an outer ST 19/38 joint on the condenser. Total height is 410mm total width is 300mm maximum O.D. is 97 mm.
Catalog No. 50-290-5819 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS33405001EA
Used for determining nitrogen in organic compounds. This assembly includes a Kjeldahl flask specifically developed for this process. Both the digestion and distillation stages can be performed in this flask. the assembly shown is the distillation stage where the amount of ammonia and hence nitrogen is established. the inlet tube allows the permanganate SOLUTIONon to be introduced after distillation. Supplied with and outer ST 29/42 joint on the flask and an outer ST 19/38 joint on the condenser. Total height is 410mm total width is 300mm maximum O.D. is 97 mm.