Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR large top desiccato1EA
Catalog No. 502908927
Encompass
These 10.5L PYREX desiccators have rugged 24/29 Standard Taper external sleeves with tooled hose connections that serves as the stopcock. They are designed so that its manipulation is not affected by the vacuum. The open position is indicated by the molded arrow on the cap. A quarter turn from this position closes the desiccator. For ease in use and improved performance it is recommended that the Standard Taper sleeve be lubricated with a silicone-based vacuum grease. The tubulation O.D. is approximately 10mm. Bowl capacity is 10 500mL. Warning Do not heat or subject to pressure. Rated 1- atmosphere vacuum.
Catalog No. 50-290-8927 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS31212501EA
These 10.5L PYREX desiccators have rugged 24/29 Standard Taper external sleeves with tooled hose connections that serves as the stopcock. They are designed so that its manipulation is not affected by the vacuum. The open position is indicated by the molded arrow on the cap. A quarter turn from this position closes the desiccator. For ease in use and improved performance it is recommended that the Standard Taper sleeve be lubricated with a silicone-based vacuum grease. The tubulation O.D. is approximately 10mm. Bowl capacity is 10 500mL. Warning Do not heat or subject to pressure. Rated 1- atmosphere vacuum.