Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR low actinic media s
Catalog No. 502900494
Encompass
Heavy-duty low actinic amber glass bottles protect light-sensitive materials. Include linerless one-piece autoclavable orange PP plug-seal caps (size GL 45) with drip-free pouring rings. Permanent white enamel graduations and marking spots.
Catalog No. 50-290-0494 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS513952504EA
Heavy-duty low actinic amber glass bottles protect light-sensitive materials. Include linerless one-piece autoclavable orange PP plug-seal caps (size GL 45) with drip-free pouring rings. Permanent white enamel graduations and marking spots.