Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR low form culture fl1EA
Catalog No. 502905840
Encompass
This narrow mouth flask is designed for culturing organisms requiring a large surface area to volume ratio. The neck O.D. is 38 mm.
Catalog No. 50-290-5840 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS44222XL1EA
This narrow mouth flask is designed for culturing organisms requiring a large surface area to volume ratio. The neck O.D. is 38 mm.