Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR petri dishes comple
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS316015012EA
Encompass
PYREX(R) petri dishes are made of clear borosilicate glass.They can withstand repeated sterilization (wet or dry) and are chemically resistant.Beaded edges provide greater mechanical strength and reduce capillary action of condensed moisture. Top marked in blue enamel bottom in white enables easier sorting.Bottoms include a triangular enamel reference point for serial dilutions.
Catalog No. 50-290-5847