Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR petri dishes comple
Catalog No. 502914674
Encompass
PYREX(R) petri dishes are made of clear borosilicate glass.They can withstand repeated sterilization (wet or dry) and are chemically resistant.Beaded edges provide greater mechanical strength and reduce capillary action of condensed moisture. Top marked in blue enamel bottom in white enables easier sorting.Bottoms include a triangular enamel reference point for serial dilutions.
Catalog No. 50-291-4674 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS31606072EA
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PYREX(R) petri dishes are made of clear borosilicate glass.They can withstand repeated sterilization (wet or dry) and are chemically resistant.Beaded edges provide greater mechanical strength and reduce capillary action of condensed moisture. Top marked in blue enamel bottom in white enables easier sorting.Bottoms include a triangular enamel reference point for serial dilutions.