Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR resin reaction kett1EA
Catalog No. 502908727
Encompass
Designed for viscous reaction materials. 500 and 1 000 mL kettles have interchangeable covers with four openings (24/40 joints). 2 000 3 000 and 4 000 mL kettles have three openings with 24/40 joints and one with a 34/45 joint. Flanges of covers and bottoms are finely ground for a tight seal. Order clamp separately.
Catalog No. 50-290-8727 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS69471L1EA
Designed for viscous reaction materials. 500 and 1 000 mL kettles have interchangeable covers with four openings (24/40 joints). 2 000 3 000 and 4 000 mL kettles have three openings with 24/40 joints and one with a 34/45 joint. Flanges of covers and bottoms are finely ground for a tight seal. Order clamp separately.